missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgA Mouse Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of mouse IgA.
£145.00 - £460.00
Specifications
For Use With (Application) | ELISA |
---|---|
Sample Size | 10 μL (prediluted 1:10,000) |
Synonym | Immunoglobulin A |
Target | Mouse |
Sample Type | Supernatant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12746867
|
Invitrogen™
88-50450-22 |
2 x 96 Tests |
£145.00
Each |
|
|||||
15590897
|
Invitrogen™
88-50450-86 |
10 x 96 Tests |
£330.00
Pack of 10 |
|
|||||
15570897
|
Invitrogen™
88-50450-76 |
20 x 96 Tests |
£460.00
Pack of 20 |
|
|||||
Description
Mouse IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
ELISA | |
Immunoglobulin A | |
Supernatant | |
2°C to 8°C | |
HRP | |
Mouse | |
HRP | |
Plasma, 10 μL|Serum, 10 μL|Supernatant, 10 μL | |
24 hr. 30 min. |
10 μL (prediluted 1:10,000) | |
Mouse | |
0.39 ng/mL | |
0.39-25 ng/mL | |
ELISA Kit | |
Colorimetric Microplate Reader | |
RUO | |
1 hr. 45 min. |
For Research Use Only