missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IFN-gamma Platinum ELISA Kit

For the quantitative detection of mouse IFN-gamma.

Brand:  Invitrogen™ BMS606TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15532017

  • £1565.85 / Pack of 10


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IFN-gamma
Biotin-Conjugate anti-mouse IFN-gamma monoclonal antibody
Streptavidin-HRP
Mouse IFN-gamma Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum
Mouse
Immune interferon, IFG
ELISA
5.3pg/mL
50μL
IFN gamma
Videos

For Research Use Only