missing translation for 'onlineSavingsMsg'
Learn More

eBioscience™ Mouse IFN gamma ELISPOT ELISA Ready-SET-Go!™ Kit

This ELISPOT reagent set is pre-titrated for optimal spot development.

Brand:  Affymetrix eBioscience 88-7384-21

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531137

  • £152.25 / Pack of 2


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Alert:

Contains Azide

Disclaimers:

For Research Use Only


Includes: Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
ELISA/ELISPOT Coating Buffer This Ready-Set-Go! ELISPOT Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISPOT Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20 and filter with 0.22 uM).
5X ELISA/ELISPOT Diluent .
Enzyme. Pre-titrated Avidin-HRP
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and enzyme

Description

Description

Mouse IFN gamma ELISPOT Ready-SET-Go! Reagent set contains the necessary reagents for performing enzyme linked immunosorbent spot (ELISPOT) assays for high resolution frequency analysis of IFNγ-secreting cells.
Specifications

Specifications

ELISPOT
Mouse
IFN gamma
2 x 96 tests
2° to 8°C
Interferon gamma
Videos