missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Granzyme B ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of Mouse Granzyme B protein levels in serum, plasma, and tissue culture supernant samples.

Brand:  Invitrogen™ 88-8022-22

Code : Z2

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 15122175

  • £140.02 / 192 tests


EU Stock 1
Add to basket

Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution

Description

Description

Mouse Granzyme B ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA).
Specifications

Specifications

ELISA
40pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
GrzB, GzB
2 x 96 tests
Mouse
Granzyme B
Videos

For Research Use Only