Learn More
Annexin V, Invitrogen™
Recombinant Protein
Brand:  Affymetrix eBioscience BMS306-1MG
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15561907
  • £1004.00 / 1mg
Estimated Shipment date
from Supplier 05-03-2020
Description
Description
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa. Description: Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells. In healthy cells, PS is predominantly located along the cytosolic side of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution in the phospholipid bilayer and translocates to the extracellular membrane, which is detectable with fluorescently labeled Annexin V. In early stages of apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI) and 7-AAD, therefore cells which display only Annexin V staining (PI/7-AAD negative) are in early stages of apoptosis. During late-stage apoptosis, loss of cell membrane integrity allows Annexin V binding to cytosolic PS, as well as cell uptake of PI and 7-AAD. Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry. Annexin V has also been identified as an anticoagulant protein in the blood coagulation cascade, by acting as an inhibitor of prothrombin activation. The presence of antibodies to Annexin V is associated with systemic lupus erythematosus (SLE), recurrent spontaneous abortions and systemic sclerosis (SSc). Purity: Annexin V is >89% pure as demonstrated by SDS-gel electrophoresis and reverse phase HPLC. Reconstitiution Instructions: We recommend a quick spin followed by reconstitution in 1 ml distilled water (incubate at 30°C for 5 minutes). Reconstituted solution contains 20 mM phosphate buffer (pH 7.0), 0.02% Tween 80, 30 mM Arginine HCL.
Specifications
Specifications
P08758, P14668, P48036
Lyophilized
308
Annexin V
SDS-gel electrophoresis and reverse phase HPLC
1mg
4°C
E. Coli
Annexin V
Control, Flow Cytometry
Recombinant Protein
Unlabeled
E. coli
Control, Flow Cytometry
20mM sodium phosphate with 0.02% Tween 80, 130mM arginine and no preservative; pH 7.0
35.8kDa
7.0
Affinity chromatography
E. coli
RUO
Recombinant
ANXA5
Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS).
All
All
Recombinant
Videos
Documents