missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse GM-CSF Quantikine ELISA Kit, R&D Systems™

Mouse GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ MGM00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442166

  • £645.09 / 192 tests

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Mouse GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse GM-CSF in cell culture supernates (50 ul), serum (50 ul).
Specifications

Specifications

7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
12981
1 Kit
Cell Culture Supernates (50uL), Serum (50uL)
Mouse
Sufficient materials to run ELISAs on at least two 96-well plates
HRP
ELISA
2 Plates
5.8pg/mL
Cell Culture Supernates (50μL), Serum (50μL)
Natural and recombinant mouse GM-CSF
Videos