missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse GM-CSF Quantikine ELISA Kit, R&D Systems™

Mouse GM-CSF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SMGM00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442816

  • £2222.64 / Pack of 6

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Mouse GM-CSF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse GM-CSF in cell culture supernates (50 ul), serum (50 ul).
Specifications

Specifications

1 Pack
Mouse
HRP
ELISA
6 Plates
Cell Culture Supernates (50uL),Serum (50uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Serum
7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
12981
5.8pg/mL
Natural and recombinant mouse GM-CSF
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents