missing translation for 'onlineSavingsMsg'
Learn More

Mouse Flt3 Ligand, Recombinant Protein, Carrier-Free, Invitrogen™

Recombinant Protein

Brand:  Affymetrix eBioscience 34-8001-85

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15588726

  • £1088.00 / 500µg

Estimated Shipment
25-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Flt3 (FMS-related tyrosine kinase 3) Ligand is a growth factor that stimulates the growth and differentiation of early hematopoetic progenitor cells. Flt3 Ligand alone exerts weak biological effects, and instead synergizes with other colony-stimulating factors to induce the proliferation and differentiation of cells of the myeloid and lymphoid lineages. Recent studies have suggested that Flt3 Ligand plays an important role in the development and expansion of dendritic cells in vivo, and that it may influence NK cell survival by this mechanism.Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform. Description: Flt3 (FMS-related tyrosine kinase 3) Ligand is a growth factor that stimulates the growth and differentiation of early hematopoetic progenitor cells. Flt3 Ligand alone exerts weak biological effects, and instead synergizes with other colony-stimulating factors to induce the proliferation and differentiation of cells of the myeloid and lymphoid lineages. Recent studies have suggested that Flt3 Ligand plays an important role in the development and expansion of dendritic cells in vivo, and that it may influence NK cell survival by this mechanism. Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform. Applications Reported: Mouse Flt3 Ligand Recombinant Protein Carrier-Free is biologically active. Applications Tested: The ED50 of this protein, as measured by IL-6 induction in M1 cells, is 50-250ng/mL in the presence of 1ng/mL Mouse LIF Recombinant Protein. This corresponds to a specific activity of 2 x 10e4 - 4 x 10e3 Units/mg. Source: E. coli expressed amino acids Thr28-Gln189 accession number NM_013520. Bioactivity: The ED50 of this protein, as measured by IL-6 induction in M1 cells, is 50-250ng/mL in the presence of 1ng/mL Mouse LIF Recombinant Protein. This corresponds to a specific activity of 2 x 10e4 - 4 x 10e3 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >97% as determined by SDS-PAGE. Molecular Weight: 20.7 kDa. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment.
Specifications

Specifications

P49772
Control, Functional Assay
Control, Functional Assay
glycine buffered saline with no preservative; pH 3.0
20.7kDa
3.0
Affinity chromatography
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
Flt3l
Recombinant Protein, Carrier-Free
Unlabeled
E. coli
Recombinant
0.5mg/mL
Liquid
14256
Mouse Flt3 Ligand (Flt3L) Carrier-Free
SDS-PAGE
500μg
-80°C
E. coli expressed amino acids Thr28-Gln189 accession number NM_013520
Mouse Flt3 Ligand (Flt3L) Carrier-Free
The ED50 of this protein, as measured by IL-6 induction in M1 cells, is 50-250ng/mL in the presence of 1ng/mL Mouse LIF Recombinant Protein. This corresponds to a specific activity of 2 x 104 - 4 x 103 Units/mg.
Mouse
Ms
Recombinant
Videos
Documents