missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL3/MIP-1 alpha DuoSet, R&D Systems™

Mouse CCL3/MIP-1 alpha DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DY450

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13433765

  • £490.98 / Pack of 15


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Description

Description

The Mouse CCL3/MIP-1 alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL3/MIP-1 alpha.
Specifications

Specifications

ELISA
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
7.81 to 500 pg/mL
15 96-well plates
Biotin
20302
Mouse
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Videos
Documents