Learn More
Invitrogen™ MCP-1/CCL2 Mouse Uncoated ELISA Kit
Uncoated ELISA Kit
Brand: Invitrogen™ 88-7391-77
349.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
The Mouse Monocyte Chemoattractant Protein-1/CCL2 (MCP-1) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse MCP-1. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
MCP-1 (Monocyte chemotactic protein-1, CCL2) has a predicted molecular weight of approximately 11kDa, however, it may run at a higher molecular weight due to glycosylation. MCP-1 is a member of the cytokine family and plays an important role in immunoregulation and inflammatory processes. MCP-1 signals through G protein-coupled receptors, CCR2 and CCR4, to recruit memory T cells, monocytes and dendritic cells. MCP-1 is linked with tumor progression, angiogenesis, and displays selective chemotactic activity towards monocytes and basophils but not neutrophils or eosinophils. MCP-1 is produced by a variety of cell types and is a potent chemoattractant for monocytes, memory T lymphocytes, and natural killer (NK) cells. Further, MCP-1 is indicated in the etiology of several immune disorders, and is upregulated during infection and inflammation. While MCP-1 is a potent basophil activator, the related protein MCP-2 stimulates both eosinophils and basophils. MCP-3 has been shown to have the broadest range of influence. MCP-1 has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis or atherosclerosis.Specifications
P10148 | |
15 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
AI553453,CCP-1/C11,CCP1,Ctla-1,Ctla1,GZB | |
HRP | |
RUO | |
2°C to 8°C | |
Mouse | |
24 hr. 30 min. |
15-2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
20296 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
20 x 96 Tests | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Monocyte chemotactic protein 1 | |
1 hr. 45 min. |
For Research Use Only