missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Mouse anti-Human, Clone: 26503, R&D Systems™
Mouse Monoclonal Antibody has been used in 53 publications
POA
Specifications
Antigen | VEGF |
---|---|
Clone | 26503 |
Applications | Western Blot, Neutralization, ELISA |
Conjugate | Unconjugated |
Form | Supernatant |
Description
VEGF Monoclonal specifically detects VEGF in Human, Primate samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
VEGF | |
Western Blot, Neutralization, ELISA | |
Supernatant | |
RUO | |
7422 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
26503 | |
Unconjugated | |
Mouse | |
Human, Primate | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 Ala27-Arg191 Accession # NP_001165097.1 | |
Primary |