Learn More
SUMO2, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against partial recombinant SUMO2.
Brand: Abnova MAB8754.100uL
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last two amino acids of the carboxy-terminus have been cleaved off. Numerous pseudogenes have been reported for this gene. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
SUMO2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:250) Western Blot (1:250-1:500) Immunofluorescence (1:250-1:500) The optimal working dilution should be determined by the end user. | |
SUMO2 | |
SUMO2 | |
Recombinant protein corresponding to amino acids 1-93 of human SUMO2. | |
RUO | |
6613 | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Mouse monoclonal antibody raised against partial recombinant SUMO2. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
HSMT3/MGC117191/SMT3B/SMT3H2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |