missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOM3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MYOM3.
Brand: Abnova H00127294-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: GPYFERPLQWKVTEDCQVQLTCKVTNTKKETRFQWFFQRAEMPDGQYDPETGTGLLCIEELSKKDKGIYRAMVSDDRGEDDTILDLTGDALDAIFTELGRIGSpecifications
MYOM3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MYOM3. | |
MYOM3 | |
FLJ35961/MGC71483/RP11-293P20.1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_152372 | |
MYOM3 | |
MYOM3 (NP_689585, 854 a.a. ∽ 955 a.a) partial recombinant protein with GST tag. | |
RUO | |
127294 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only