missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Mouse anti-Human, Clone: 8516, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | IL-1 beta/IL-1F2 |
---|---|
Clone | 8516 |
Applications | Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF |
Conjugate | Unconjugated |
Form | Purified |
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
IL-1 beta/IL-1F2 | |
Western Blot, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
Purified | |
RUO | |
3553 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
8516 | |
Unconjugated | |
Mouse | |
Human | |
E. coli-derived recombinant human IL-1 beta/IL-1F2 aa 117-269 Accession # P01584 | |
Primary |