missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hepatitis B virus presurface antigen, Mouse, Clone: S 26, Abnova™
Mouse monoclonal antibody raised against native hepatitis B virus presurface antigen.
Brand: Abnova MAB5399.100ug
Additional Details : Weight : 0.00010kg
Specifications
Hepatitis B virus presurface antigen | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
S 26 | |
Mouse monoclonal antibody raised against native hepatitis B virus pre-surface antigen. | |
In serum | |
Native HBsAg from human sera pool. | |
RUO | |
HBsAg pre-S antigen. Recognizes a.a. 132-a.a. 137 of pre-S (adw nomenclature). | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |