missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIAPH3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DIAPH3.
Brand: Abnova H00081624-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: PDILNFVDDLEPLDKASKVSVETLEKNLRQMGRQLQQLEKELETFPPPEDLHDKFVTKMSRFVISAKEQYETLSKLHENMEKLYQSIIGYYAIDVKKVSpecifications
DIAPH3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant DIAPH3. | |
DIAPH3 | |
DKFZp434C0931/DKFZp686A13178/DRF3/Dia2/FLJ34705/diap3 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_030932 | |
DIAPH3 | |
DIAPH3 (NP_112194, 632 a.a. to 729 a.a) partial recombinant protein with GST tag. | |
RUO | |
81624 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only