Learn More
COL9A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COL9A1.
Brand: Abnova H00001297-A01.50uL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of the three alpha chains of type IX collagen, which is a minor (5-20%) collagen component of hyaline cartilage. Type IX collagen is usually found in tissues containing type II collagen, a fibrillar collagen. Studies in knockout mice have shown that synthesis of the alpha 1 chain is essential for assembly of type IX collagen molecules, a heterotrimeric molecule, and that lack of type IX collagen is associated with early onset osteoarthritis. Mutations in this gene are associated with osteoarthritis in humans, with multiple epiphyseal dysplasia, 6, a form of chondrodysplasia, and with Stickler syndrome, a disease characterized by ophthalmic, orofacial, articular, and auditory defects. Two transcript variants that encode different isoforms have been identified for this gene. [provided by RefSeq
Sequence: AVKRRPRFPVNSNSNGGNELCPKIRIGQDDLPGFDLISQFQVDKAASRRAIQRVVGSATLQVAYKLGNNVDFRIPTRNLYPSGLPEEYSFLTTFRMTGSTLKKNWNIWQSpecifications
COL9A1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant COL9A1. | |
COL9A1 | |
DJ149L1.1.2/EDM6/FLJ40263/MED | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001851 | |
COL9A1 | |
COL9A1 (NP_001842, 24 a.a. to 132 a.a) partial recombinant protein with GST tag. | |
RUO | |
1297 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only