Learn More
CD3G, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CD3G.
Brand: Abnova H00000917-A01.50uL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in this gene are associated with T cell immunodeficiency. [provided by RefSeq
Sequence: QSIKGNHLVKVYDYQEDGSVLLTCDAEAKNITWFKDGKMIGFLTEDKKKWNLGSNAKDPRGMYQCKGSQNKSKPLQVYYRMCQNCIELNSpecifications
CD3G | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CD3G. | |
CD3G | |
CD3-GAMMA/FLJ17620/FLJ17664/FLJ79544/FLJ94613/MGC138597/T3G | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000073 | |
CD3G | |
CD3G (NP_000064, 23 a.a. to 111 a.a) partial recombinant protein with GST tag. | |
RUO | |
917 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only