missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-CALR, Clone: 1G6A7, Abnova™
Brand: Abnova MAB10459.100uL
Additional Details : Weight : 0.02000kg
Specifications
Calreticulin | |
Monoclonal | |
Unconjugated | |
Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
CALR | |
CALR | |
A synthetic peptide (conjugated with KLH) corresponding to human Calreticulin. | |
RUO | |
811 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
1G6A7 | |
Mouse monoclonal antibody raised against synthetic peptide CALR. | |
In ascitic (0.03% sodium azide) | |
CRT/FLJ26680/RO/SSA/cC1qR | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |