Learn More
BMPR1A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant BMPR1A.
Brand: Abnova H00000657-A03.50uL
Additional Details : Weight : 0.00010kg
Description
The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55kDa and type II receptors of about 70-80kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. [provided by RefSeq
Sequence: MPQLYIYIRLLGAYLFIISRVQGQNLDSMLHGTGMKSDSDQKKSENGVTLAPEDTLPFLKCYCSGHCPDDAINNTCITNGHCFAIIEEDDQGETTLASGCMKYEGSGFQCKDSPKAQLRRTIECCRTNLCNQYLQPTLPPVVIGPFFDGSIRWLVLLISMAVCIIAMIIFSSCFCYKHYCKSISSRRRYNRDLEQDEAFIPVGESLKDLVDQSQSSGSGSGLPLLVQRTIAKQIQMVRQVGKGRYGEVWMGKWRGEKVAVKVFFTTEEASWFRETEIYQTVLMRHENILGFIAADIKGTGSWTQLYLITDYHENGSLYDFLKCATLDTRALLKLAYSAACGLCHLHTEIYGTQGKPAIAHRDLKSKNILIKKNGSCCIADLGLAVKFNSDTNEVDVPLNTRVGTKRYMAPEVLDESLNKNHFQPYIMADIYSFGLIIWEMARRCITGGIVEEYQLPYYNMVPSDPSYEDMREVVCVKRLRPIVSNRWNSDECLRAVLKLMSECWAHNPASRLTALRIKKTLAKMVESQDVKISpecifications
BMPR1A | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant BMPR1A. | |
BMPR1A | |
10q23del/ACVRLK3/ALK3/CD292 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC028383 | |
BMPR1A | |
BMPR1A (AAH28383, 1 a.a. to 532 a.a) full-length recombinant protein with GST tag. | |
RUO | |
657 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only