missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Adiponectin/Acrp30 Quantikine ELISA Kit, R&D Systems™

Mouse Adiponectin/Acrp30 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ MRP300

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442216

  • £545.37 / 96 tests

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Mouse Adiponectin/Acrp30 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Adiponectin/Acrp30 in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications

Specifications

0.16 - 10ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
11450
1 Plate
0.007ng/mL
Cell Culture Supernates (50μL), Tissue Homogenates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), Tissue Homogenates (50uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL)
Mouse
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents