missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morin hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | 300.0°C |
Color | Yellow to Brown |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 50 g |
Beilstein | 18, 239 |
Merck Index | 14, 6269 |
Solubility Information | Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water, soluble in organic solvents |
Formula Weight | 302.24 |
Physical Form | Powder |
Show More |
Safety and Handling
Product Identifier
- Morin hydrate
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'