missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, Ultrapure Bioreagent, J.T. Baker™
627.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
- For Molecular Biology Applications
Specifications
Flash Point | 116.00°C |
Packaging | Poly Bottle |
Quantity | 500 g |
IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
Formula Weight | 209.26 |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | MOPS |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'