missing translation for 'onlineSavingsMsg'
Learn More

MOPS, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Catalog Number p-7082835
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-[N-Morpholino]propane sulfonic Acid
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
C1COCCN1CCCS(=O)(=O)O
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-[N-Morpholino]propane sulfonic Acid
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
C1COCCN1CCCS(=O)(=O)O
Melting Point 277°C to 280°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
Flash Point 110°C
Linear Formula HO3S(CH2)3(C4H8NO)
Packaging Plastic bottle
Beilstein 1106776
Formula Weight 209.26
Percent Purity ≥98.5%
Assay Dry basis
Chemical Name or Material MOPS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'