missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elgas™ Monoplug PLUG36860 230V 50/60Hz 3.5mm Elga
Brand: Elgas™ CEPLGU0125
Code : 50
Additional Details : Weight : 1.00000kg
Brand: Elgas™ CEPLGU0125
Code : 50
Additional Details : Weight : 1.00000kg