missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl adipate, 97%, Thermo Scientific Chemicals

Catalog Number p-170058
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
627-91-8
C7H12O4
160.17
MFCD00004418
UOBSVARXACCLLH-UHFFFAOYSA-N
monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
12328
CHEBI:70855
6-methoxy-6-oxohexanoic acid
COC(=O)CCCCC(O)=O
This item is not returnable. View return policy
627-91-8
C7H12O4
160.17
MFCD00004418
UOBSVARXACCLLH-UHFFFAOYSA-N
monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
12328
CHEBI:70855
6-methoxy-6-oxohexanoic acid
COC(=O)CCCCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 9.0°C
Color Colorless
Density 1.0800g/mL
Boiling Point 162.0°C (10.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HO2C(CH2)4CO2CH3
Refractive Index 1.4390 to 1.4420
Quantity 5 g
Beilstein 02, III, 1711
Specific Gravity 1.08
Formula Weight 160.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Monomethyl adipate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Monomethyl adipate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'