missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl 5-nitroisophthalate, 98%, Acros Organics™

Brand:  Acros Organics 217750250

Code : 29

Additional Details : CAS Number : 1955-46-0 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10246940

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

mono-Methyl 5-nitroisophthalate
97.5
98%
C9H7NO6
MFCD00009793
1955-46-0
100.0
98%
225.16

Specifications

mono-Methyl 5-nitroisophthalate
97.5
Authentic
98%
C9H7NO6
MFCD00009793
Solubility in water: insoluble.
Slightly yellow
1955-46-0
100.0
Fine Crystalline Powder
98%
225.16
Glass bottle
181.0° to 185.0°C
25g
Videos
SDS