Learn More
Invitrogen™ E-selectin (Soluble) Monkey ELISA Kit
For the quantitative detection of monkey sE-selectin.
Brand: Invitrogen™ BMS649TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sE-selectin
HRP-Conjugate anti-monkey sE-selectin monoclonal antibody
Monkey sE-selectin Standard lyophilized, 120 U/ml upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
The Monkey Endothelial Leukocyte Adhesion Molecule (Mk e-Selectin/ELAM-1) ELISA quantitates Mk ELAM-1 in baboon serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Mk ELAM-1. Principle of the method The Monkey ELAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
E-selectin (CD62E, ELAM-1, and LECAM-1) is a 115 kD type I membrane protein and a member of the selectin family. E-selectin is highly expressed on activated endothelial cells (IL-2, TNF- alpha, other cytokines can increase expression) and can also be expressed on endothelial cells in the skin, bone marrow and placenta. E-selectin is involved in tethering and leucocyte rolling on activated endothelium at inflammatory sites and may also play a role in tumor metastasis and angiogenesis. E-selectin binds to both Siayl Lewis X (CD15s) and PSGL-1 (CD162). The HCD62E antibody has been shown to recognize human E-selectin and to be useful for flow cytometry. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.Specifications
1.9-60 U/mL | |
HRP | |
Serum, Supernatant | |
ELISA | |
101009590 | |
10% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 20 μL; Supernatant, 20 μL | |
ELAM1, endothelial adhesion molecule 1, CD62E | |
1 hr. 20 min. |
0.4 U/mL | |
ELISA Kit | |
Monkey | |
Colorimetric Microplate Reader | |
SELE | |
5% | |
HRP | |
RUO | |
2°C to 8°C | |
Baboon | |
2 hr. 10 min. |
For Research Use Only