Learn More
Invitrogen™ IL-6 Monkey ELISA Kit
For the quantitative detection of monkey IL-6.
£451.00 - £2805.00
Specifications
Accession Number | P79341 |
---|---|
Assay Range | 1.9-120 U/mL |
Assay Sensitivity | 1.3 U/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-6
Biotin-Conjugate anti-monkey IL-6 monoclonal antibody
Streptavidin-HRP
Monkey IL-6 Standard lyophilized, 240 U/ml upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15572077
|
Invitrogen™
BMS641-2TEN |
10 x 96 Tests |
£2805.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15562077
|
Invitrogen™
BMS641-2 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
The Monkey Interleukin 6 (Mk IL-6) ELISA quantitates Mk IL-6 in monkey serum or cell culture medium. The assay will exclusively recognize both natural and recombinant monkey IL-6. Principle of the method The Monkey IL-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P79341 | |
1.3 U/mL | |
ELISA Kit | |
Monkey | |
Colorimetric Microplate Reader | |
EGK_13861 | |
5% | |
HRP | |
Serum, 50 μL; Supernatant, 50 μL | |
Interferon-beta 2, B-cell stimulatory factor 2,HGF | |
1 hr. 20 min. |
1.9-120 U/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
100381195, 102138971 | |
10% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Baboon, Cynomolgus Monkey | |
3 hr. 10 min. |
For Research Use Only