Learn More
Invitrogen™ IL-2 Monkey ELISA Kit
For the quantitative detection of monkey IL-2.
Brand: Invitrogen™ BMS643-2TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-2
Description
The Monkey Interleukin 2 (Mk IL-2) ELISA quantitates Mk IL-2 in monkey serum or cell culture medium. The assay will exclusively recognize both natural and recombinant monkey IL-2. Principle of the method The Monkey IL-2 solid-phase sandwich E
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells,differentiation of B cells,natural killer cells,monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R),a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic,which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
Q29615,Q865Y1 | |
9.1 U/mL | |
Amniotic Fluid, Plasma, Serum, Supernatant, Urine, Whole Blood | |
ELISA | |
100126754,102129830 | |
5% | |
HRP | |
RUO | |
2°C to 8°C | |
Baboon,Cynomolgus Monkey | |
3 hr. 30 min. |
18.8-1,200 U/mL | |
ELISA Kit | |
Monkey | |
Colorimetric Microplate Reader | |
IL2 | |
7% | |
10 x 96 Tests | |
Amniotic Fluid, 50 μL; Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL; Urine,50 μL; Whole Blood,50 μL | |
Aldesleukin, T cell growth factor, TCGF | |
1 hr. 20 min. |