Learn More
Monel|r 400 wire, 0.50mm (0.02in) dia, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045066.TC
Code : NEW
Additional Details : Weight : 0.22700kg
Description
Alloy 400 is widely used in many fields, especially marine and chemical processing. Typical applications are valves and pumps; pump and propeller shafts; marine fixtures and fasteners; electrical and electronic components; springs; chemical processing equipment; gasoline and fresh water tanks; crude petroleum stills, process vessels and piping; boiler feedwater heaters and other heat exchangers; and deaerating heaters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Monel™ 400 wire | |
MFCD09910536 | |
Insoluble in water. | |
Wire |
0.50mm (0.02 in.) dia. | |
227 g | |
≈1.7g/m |
RUO – Research Use Only