missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Monel™ 400 gauze, 80 mesh woven from 0.14mm (0.0055 in.) dia. wire

Brand:  Alfa Aesar™ 045207.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15416825

  • £18.90 / Each


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Monel™ 400 gauze
80 mesh woven from 0.14mm (0.0055 in.) dia. wire

Specifications

Monel™ 400 gauze
80 Mesh
150 x 150mm
80 mesh woven from 0.14mm (0.0055 in.) dia. wire
Videos
SDS