missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Monel™ 400 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire

Brand:  Alfa Aesar™ 045163.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150 x 150mm

 View more versions of this product

Product Code. 15406795

  • £17.10 / Each


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Monel™ 400 gauze
40 mesh woven from 0.25mm (0.01 in.) dia. wire

Specifications

Monel™ 400 gauze
40 Mesh
150 x 150mm
40 mesh woven from 0.25mm (0.01 in.) dia. wire
Videos
SDS