missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Monel™ 400 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire

Brand:  Alfa Aesar™ 045163.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300 x 300mm

 View more versions of this product

Product Code. 15416795

  • £55.90 / Each


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Monel™ 400 gauze
40 mesh woven from 0.25mm (0.01 in.) dia. wire

Specifications

Monel™ 400 gauze
40 Mesh
300 x 300mm
40 mesh woven from 0.25mm (0.01 in.) dia. wire
Videos
SDS