missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum wire, 0.013mm (0.00052 in.) dia., hard, 99.8% (metals basis)

Microelectronics, high temp

Brand:  Alfa Aesar™ 040669.G6

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg

Quantity 10m

 View more versions of this product

Product Code. 11378598

  • £415.80 / 10m


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Molybdenum wire
7439-98-7
0.013mm (0.00052 in.) dia., Hard
MFCD00003465

Specifications

Molybdenum wire
10m
7439-98-7
99.8%
MFCD00003465
≈0.0014g/m
0.013mm (0.00052 in.) dia., Hard
Wire
(metals basis)
Videos
SDS