missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum wire, 0.013mm (0.00052 in.) dia., hard, 99.8% (metals basis)

Microelectronics, high temp

Brand:  Alfa Aesar™ 040669.G5

UNSPSC : 12141727

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg

Quantity 5m

 View more versions of this product

Product Code. 11358598

  • £255.57 / 5m


Estimated Shipment date
from Supplier 13-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Molybdenum wire
7439-98-7
0.013mm (0.00052 in.) dia., Hard
MFCD00003465

Specifications

Molybdenum wire
5m
7439-98-7
99.8%
MFCD00003465
≈0.0014g/m
0.013mm (0.00052 in.) dia., Hard
Wire
(metals basis)
Documentation

Technical Documents