missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog Number 11314217
Click to view available options
Quantity:
100 g
500 g
1313-27-5
MoO3
143.95
MFCD00003469
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O
1313-27-5
MoO3
143.95
MFCD00003469
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O

Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in oil industr. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst
TRUSTED_SUSTAINABILITY
Boiling Point 1155°C (sublimation)
Melting Point 795°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5%
UN Number UN3288
Merck Index 14,6239
Solubility Information Slightly soluble in water; Soluble in concentrated mineral acids,alkali hydroxide solutions.
Formula Weight 143.94
Percent Purity ≤99.5%
Grade ACS
Density 4.692 g/mL
Chemical Name or Material Molybdenum oxide, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'