missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(VI) oxide, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1155.0°C |
Melting Point | 795.0°C |
Quantity | 500 g |
Assay Percent Range | 99.5+% |
Linear Formula | MoO3 |
Merck Index | 15,6325 |
Solubility Information | Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in conc. mineral acids, sol. of alkali, hydroxides and ammonia, soluble in potassium bitartrate |
Formula Weight | 143.94 |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Show More |
Safety and Handling
Product Identifier
- Molybdenum(VI) oxide
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'