Learn More
Thermo Scientific Alfa Aesar Molybdenum Thinfoil, 0.0025mm (0.0001 in.) thick, 99.9% (metals basis)
Brand: Thermo Scientific Alfa Aesar 040710.FF
Molybdenum Thinfoil, 0.0025mm (0.0001 in.) thick, 99.9% (metals basis)
Code : NEW
Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg