missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Brand:  Alfa Aesar™ 041078.KS

Code : NEW

Additional Details : CAS Number : 12136-78-6 Weight : 0.01000kg

Product Code. 15414845

  • £261.82 / Each


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Specifications

Molybdenum silicide sputtering target
76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
6.31g/mL
99.5%
152.13
Odorless
1870° to 2030°C
1 each
12136-78-6
Target
(metals basis)
MFCD00151535
Dark gray
Videos
SDS