missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, 99.95% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 010030.06

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.00500kg Transport : UN number : 3089 Chem class : 4.1 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 15435915

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Molybdenum
Powder
MFCD00003465
7439-98-7
99.95% (Metals basis)

Specifications

Molybdenum
7439-98-7
Powder
MFCD00003465
Gray
5g
gray powder
99.95% (Metals basis)
Odorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Molybdenum powder

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.

SDS