missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, APS 3-7 micron, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 15445915
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance gray powder
Density 10.22g/cm3 at 20°C
Assay Percent Range 99.95% (Metals basis)
Quantity 50 g
Odor Odorless
Form Powder
Chemical Name or Material Molybdenum
compliance-icons
Product Identifier
  • Molybdenum powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use fire-fighting equipment on basis class D to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Molybdenum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'