missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum powder, -250 mesh, 99.9% (metals basis)

Brand:  Alfa Aesar™ 000932.18

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.05000kg Transport : UN number : 3089 Chem class : 1 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 15495905

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Molybdenum powder
7439-98-7
MFCD00003465
UN3089

Specifications

Molybdenum powder
7439-98-7
99.9%
MFCD00003465
−250 Mesh
Gray
50g
Powder
(metals basis)
UN3089
Odorless
Videos
SDS