missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(II) acetate dimer, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Molybdenum(II) acetate dimer is generally used as an intermediate compound in a process to form other quadruply bonded molybdenum compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Slightly soluble in organic solvents. Insoluble in water. |
Formula Weight | 428.06 |
Physical Form | Crystalline |
Chemical Name or Material | Molybdenum(II) acetate dimer |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'