missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum gauze, 50 mesh woven from 0.0509mm (0.002 in.) dia. wire

Brand:  Alfa Aesar™ 040911.FL

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg

Quantity 75x75mm

 View more versions of this product

Product Code. 11315197

  • £118.10 / Each


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

Molybdenum gauze
7439-98-7
Woven from 0.0509mm (0.002 in.) dia. wire
MFCD00003465

Specifications

Molybdenum gauze
Woven from 0.0509mm (0.002 in.) dia. wire
Wire Cloth
50 Mesh
75x75mm
7439-98-7
MFCD00003465
Videos
SDS