missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum gauze, 10 mesh woven from 0.38mm (0.015mm) dia wire

Brand:  Alfa Aesar™ 046494.HB

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg

Quantity 300x300mm

 View more versions of this product

Product Code. 15477225

  • £213.00 / Each


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

Molybdenum gauze
7439-98-7
10 mesh woven from 0.38mm (0.015mm) dia. wire
MFCD00003465

Specifications

Molybdenum gauze
10 mesh woven from 0.38mm (0.015mm) dia. wire
Wire Cloth
10 Mesh
300x300mm
7439-98-7
MFCD00003465
Videos
SDS