Learn More
Molecular sieves, tantalum oxide based; mesoporous, 22 Angstrom, powder, Thermo Scientific™
Catalysis, gas adsorption, optoelectronic materials
Brand: Thermo Scientific Alfa Aesar 042612.MD
Code : NEW
Additional Details : Weight : 0.00025kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hygroscopic | |
Molecular sieves, tantalum oxide based | |
Mesoporous | |
MFCD00131613 |
250 mg | |
22Å | |
Powder | |
Ta TMSI |