missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieve, Activated, Type 3A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Specifications
Mesh Size | 8 to 12 Mesh |
Packaging | Wide Mouth PVC Coated Glass Bottle |
Specific Gravity | 1L = 2.10kg |
Quantity | 2.5 kg |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Molecular Sieve, Activated, Type 3A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'