missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Valinomycin

Commonly used as an agent to induce apoptosis and can calibrate potentiometric responses in membrane experiments involving potential-sensitive dyes

Brand:  Invitrogen™ V1644

Code : 50

Additional Details : CAS Number : 2001-95-8 Weight : 0.00450kg

Product Code. 11510816

  • £133.00 / 25mg
Estimated Shipment: 24-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • Valinomycin is a natural, lipid-soluble molecule (Mr = approximately 1110) that binds potassium ions and facilitates their transfer across lipid bilayers (i.e., it is an ionophore)
  • A macrocyclic molecule formed from 12 alternating amino acids and esters, synthesized by specific Streptomyces
  • Exhibits a 10,000-fold selectivity for K+ over Na+
  • Valinomycin is a potent antibiotic
  • K+ selective ionophore
  • Neutrally charged
  • Soluble in ethanol, DMSO, etc., but insoluble in water

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Valinomycin
Ionophore
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.