missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ Red Fluorescent Nucleic Acid Stain Sampler Kit - SYTO™ dyes 17 and 59-64 - 50 μL each

Product Code. 10685193
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10685193

Brand: Invitrogen™ S11340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For use with fluorescence microscopy and flow cytometry

The SYTO Red Fluorescent Nucleic Acid Stain Sampler Kit contains our collection of cell-permeant, red-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 17 and 59–64 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range 621, 622, 652, 628, 652, 657, 599 nm
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 1 Kit
Volume (Metric) 50 μL
Detection Method Fluorescence
Description SYTO™ Red Fluorescent Nucleic Acid Stain Sampler Kit - SYTO™ dyes 17 and 59-64, 50 μL each
Emission 634, 645, 678, 645, 676, 673, 619 nm
Product Line SYTO
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Stains Dimer Sampler Kit
SubCellular Localization Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.